Skip to Content
Merck
All Photos(1)

Key Documents

Z208175

Sigma-Aldrich

Rubbermaid® heavy-duty lab carts

black

Synonym(s):

heavy-duty cart, rubbermaid cart, rubbermaid carts

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$574.00

$574.00


Available to ship on23 April 2025Details



Select a Size

Change View
1 EA
$574.00

About This Item

UNSPSC Code:
41100000
NACRES:
JA.01

$574.00


Available to ship on23 April 2025Details


color

black

Looking for similar products? Visit Product Comparison Guide

General description

Duramold® construction holds up under rugged use. Seamless construction, no sharp edges. Easy to clean. Maneuverable. Optional middle shelf sold separately for models B and C.

Features and Benefits

  • 500-lb. mass capacity
  • Made for transport of supplies and heavy loads
  • Push handle provides optimum hand placement and push height for better control and safety
  • Molded-in storage compartments, holsters, hooks and notches
  • Larger work surface for oversized loads
  • Structural resin construction
  • Rounded corners protects walls and furniture
  • Bottom shelf for easy access and keeping contents secure
  • 4-in. non-marking swivel casters hold up in tough environments
  • Overall Dimensions: 39-in. L x 17.9-in. W x 33.2-in. H
  • Utility Cart, 39"L x 17-7/8"W x 33-1/4"H, 500 lb. capacity, (2) shelves, open base, polyethylene exterior, (2) fixed and (2) swivel 5" thermoplastic (TPR) casters, assembly required, black

Legal Information

Duramold is a registered trademark of Rubbermaid Commercial Products, Inc.
Rubbermaid is a registered trademark of Rubbermaid Commercial Products, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service