Z262250
KNF Laboport® solid PTFE vacuum pump
pumping speed 10 L/min, AC/DC input 115 V AC
Synonym(s):
laboratory vacuum pump, mini diaphragm vacuum pump, mini vacuum pump, vacuum pump
About This Item
Recommended Products
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
KNF
parameter
≤6 torr vacuum
10 L/min pumping speed
L × W × H
11 in. × 5 1/2 in. × 7.2 in.
weight
16 lb (7.3 kg)
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Solid PTFE headsPTFE coated diaphragm.
- Oil-free operation ensures pumped medium will stay pure.
- New multi-port valve system with Kalrez disks improves flow and reliability.
- Two stages.
- 0 mm I.D. hose barbs on ports.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service