Key Documents
Z259551
Sigma-Aldrich
Ace photochemical immersion well
Quartz
Select a Size
1 EA
$1,870.00
$1,870.00
Estimated to ship on26 March 2025
Select a Size
Change View
1 EA
$1,870.00
About This Item
UNSPSC Code:
41121800
NACRES:
NC.25
$1,870.00
Estimated to ship on26 March 2025
manufacturer/tradename
Ace Glass 787435
Related Categories
General description
Double-walled, with small diameter inner tube that extends down the annular space to insure flow of coolant from bottom of well upwards to outlet.
Compression seal inside reactor vessel allows adjustable height of well. Total length: 390 mm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)