Key Documents
Z214612
Micro photochemical reactor UV lamp
output5.5 watts
Select a Size
1 EA
$1,790.00
$1,790.00
Check Cart for Availability
Select a Size
Change View
1 EA
$1,790.00
About This Item
UNSPSC Code:
41103817
$1,790.00
Check Cart for Availability
feature
CE compliant: no
CSA approved: no
manufacturer/tradename
Ace Glass 1213208
availability
not available in EU, Canada
output
5.5 watts
General description
Cold cathode, low pressure, mercury arc, gaseous discharge lamps made of double bore quartz. Lamp power consumption is 5.5 watts, with principle output at 254 nanometers. Lamps are rated for 5000 hours of operation.
Choose from one of the most recent versions:
Certificates of Analysis (COA)