Key Documents
Z243868
Sigma-Aldrich
POWERMITE proportional on/off type-1,000W capacity heat control
EB-1C
About This Item
UNSPSC Code:
41112303
NACRES:
NB.76
Product Z243868 is not currently sold in your country. Contact Technical Service
AC/DC input
120 V AC
feature
CSA approved
availability
not available in EU
weight
1.6 lb
Related Categories
General description
Model EB-1C has a cast aluminum case with gray, baked-on finish, satin SS face plate with flush-mounted receptacle and dial plate, pilot lamp (two lamps on model EB-1C), and a 3 ft, 3-wire power cord with U.S. plug.
Used to control THERMOWELL heating mantles or other resistive heating circuits (hot plates, heat tapes, water baths, etc.). Available in solid-state or proportional on/off models. All models are fused (without circuit breakers). Each unit is calibrated to deliver a known amount of power or voltage at a given dial setting in the 15 to 70% range (off below 15% and full on above 70%). High capacity with constant power output despite large line voltage fluctuations. Does not vary voltage. 50/60 Hz.
Legal Information
POWERMITE is a trademark of Laboratory Craftsmen, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)