Key Documents
Z220108
Sigma-Aldrich
Aldrich® micro distillation head with Vigreux indents
One-piece assembly
Select a Size
1 EA
$3,110.00
$3,110.00
Check Cart for Availability
Select a Size
Change View
1 EA
$3,110.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
$3,110.00
Check Cart for Availability
Related Categories
General description
Has a vacuum-jacketed head with attached condenser, Vigreux indents and thermowell for 50 mm immersion thermometer. Joints on the one-piece are 14/20 inner joints and 10/18 outer joints. Vigreux indents run the entire length of the column and act as the packing for the column.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)