Key Documents
Z200336
Squeeze-bulb pump
internal flapper valve prevents reverse flow, pkg of 1 ea
Select a Size
1 EA
$34.00
$34.00
Check Cart for Availability
Select a Size
Change View
1 EA
$34.00
About This Item
UNSPSC Code:
40151500
NACRES:
NB.51
$34.00
Check Cart for Availability
packaging
pkg of 1 ea
General description
PE, 1 to 1 1/2 gpm. Flexible discharge tube and hose hook with tapered adapter fit 1 to 5 gal containers or cans. Internal flapper valve prevents reverse flow.
Uses: mild solvents, light oils, corrosives, dilute acid/caustics, liquid waxes, and disinfectants. Barrel: 21 in. L. Hose: 38 L × 1 in. diameter.
Choose from one of the most recent versions:
Certificates of Analysis (COA)