Key Documents
Z173568
Sigma-Aldrich
Aldrich® Luer lock adapter
female joint: ST/NS 24/40 to male Luer lock
Select a Size
1 EA
$242.00
$242.00
Check Cart for Availability
Select a Size
Change View
1 EA
$242.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$242.00
Check Cart for Availability
Joint
female joint: ST/NS24/40 to male Luer lock
Related Categories
General description
Permits fitting of standard glassware with needles for air-sensitive transfer, etc.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)