Key Documents
Z170658
Sigma-Aldrich
Aldrich® glass mercury trap
pkg of 1 ea
Select a Size
1 EA
$365.00
$365.00
Check Cart for Availability
Select a Size
Change View
1 EA
$365.00
About This Item
UNSPSC Code:
41112406
NACRES:
NB.79
$365.00
Check Cart for Availability
packaging
pkg of 1 ea
Related Categories
General description
Designed to prevent mercury from being drawn into vacuum pump.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)