Erado-Sol® liquid stain remover is a product primarily designed for removing stains on fabrics and upholstery. It is not specifically formulated for removing water stains on wood. To effectively remove water stains from wood, it is recommended to use products specifically designed for that purpose.
Z151564
Erado-Sol® liquid stain remover
weight 32 oz (964 g ), (bottle)
Select a Size
$85.50
Select a Size
About This Item
$85.50
Recommended Products
weight
32 oz (964 g ), (bottle)
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
will this remove water stains from wood
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service