Key Documents
Z125458
Ace-Thred Thermometer/Tubing Adapter
Straight, joint: ST/NS 24/40, thread I.D. 9.0-10.5 mm
Select a Size
1 EA
$104.00
$104.00
Check Cart for Availability
Select a Size
Change View
1 EA
$104.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$104.00
Check Cart for Availability
label
A
Joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass 503022
thread I.D.
9.0-10.5 mm
Related Categories
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)