Key Documents
Z120960
Syringe pressure caps
sterile
Select a Size
100 EA
$45.70
$45.70
Check Cart for Availability
Select a Size
Change View
100 EA
$45.70
About This Item
UNSPSC Code:
41121800
NACRES:
NB.03
$45.70
Check Cart for Availability
material
polyolefin
sterility
sterile
packaging
bag of 100 ea
General description
Disposable. For use with Luer lock or regular Luer syring tips, covers entire Luer skirt of syringe.
Choose from one of the most recent versions:
Certificates of Analysis (COA)