Skip to Content
Merck
All Photos(1)

Documents

Z115029

Sigma-Aldrich

Serum-finish V-Vials®

volume 0.1 mL, mouth O.D. 11 mm, diam. × H 12 mm × 32 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

material

clear glass vial

feature

closure type crimp top vial

diam. × H

12 mm × 32 mm

mouth O.D.

11 mm

volume

0.1 mL

Looking for similar products? Visit Product Comparison Guide

General description

Vials only. Autoclavable borosilicate USP Type 1 glass. Uses 11 mm aluminum seals. The 0.1mL size fits standard GC autosamplers.

Legal Information

V-Vial is a registered trademark of Wheaton Industries, Inc.

related product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service