Skip to Content
Merck
All Photos(1)

Key Documents

33892

Supelco

T-2 Toxin-13C24 solution

~25 μg/mL in acetonitrile, analytical standard

Synonym(s):

Fully 13C-labelled T-2 Toxin, U-[13C24] - T-2 Toxin

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$2,650.00

$2,650.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$2,650.00

About This Item

Empirical Formula (Hill Notation):
13C24H34O9
Molecular Weight:
490.35
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

$2,650.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~25 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

General description

T-2 Toxin-13C24 is the 13C isotope-labelled analog of mycotoxin, T-2 Toxin

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service