Key Documents
Z111554
Sigma-Aldrich
Steel vial and ball pestle for analog Wig-L-Bug® grinder/mixer
stainless steel, diam. × L 1/2 in. × 1.0 in.
Select a Size
1 EA
$196.00
$196.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$196.00
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
$196.00
Check Cart for Availability
Request a Bulk Order
material
stainless steel
availability
not available in EU
technique(s)
IR spectroscopy: suitable
diam. × L
1/2 in. × 1.0 in.
General description
For grinding and mixing hard materials. Includes 1/4 in. diam. ball pestle.
Legal Information
Wig-L-Bug is a registered trademark of Dentsply Rinn
Choose from one of the most recent versions:
Certificates of Analysis (COA)