Skip to Content
Merck
All Photos(1)

Key Documents

Z106186

Wheaton connector

13-425 to 20-400

Synonym(s):

125480

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$53.00

$53.00


Check Cart for Availability


Select a Size

Change View
1 EA
$53.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

$53.00


Check Cart for Availability

manufacturer/tradename

WHEATON®

Looking for similar products? Visit Product Comparison Guide

General description

Grease-free with screw threads to provide an inert connection between outer-threaded glass components. All wetted parts are PTFE. Dynamic vacuum of <5 μm is achieved. May be used at temperatures up to 180 °C.

Features and Benefits

  • Safely joins two exterior threaded glass components without hooks, springs or clamps
  • Screw cap configuration creates a grease-free, vacuum-tight seal
  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Available with white polypropylene or black phenolic caps
  • Phenolic connectors have a working temperature of up to 180°C
  • Autoclavable

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service