WHA9915-2504
Whatman® UNIFLO® 25 syringe filters
polyethersulfone membrane, sterile, pore size 0.45 μm, pack of 200 ea
Select a Size
$548.00
Select a Size
About This Item
$548.00
Recommended Products
material
polyethersulfone membrane
polypropylene housing
sterility
sterile
packaging
pack of 200 ea
manufacturer/tradename
Whatman 9915-2504
diam.
25 mm
pore size
0.45 μm
fitting
female inlet fitting for Luer Lock
male outlet fitting for Slip Luer
suitability
suitable for (volumes of 100mL or less; filter provides easy separation of solutions)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- 13 or 25 mm diameters
- 0.2 µm, 0.22 µm or 0.45 µm pore sizes
- Sterile or non-sterile options
- Individual printing on the filter for easy identification
- Bench top space saving packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service