WHA69712504
Whatman® GD/XP syringe filters
Nylon membrane
Synonym(s):
Z754684
Select a Size
$4,350.00
Select a Size
About This Item
$4,350.00
Recommended Products
material
Nylon membrane
polypropylene housing
sterility
non-sterile
packaging
pk of 1500
manufacturer/tradename
Whatman 6971-2504
Whatman Article No. 28420562 (US reference)
diam.
25 mm
size
volume
≤25 mL
pore size
0.45 μm
fitting
female Luer inlet (lock)
male Luer outlet
Features and Benefits
- Incorporates a two-layer polypropylene prefilter stack rather than a multi-layer glass fiber stack to minimize levels of ion extractables
- Extremely low protein binding syringe filter
- High loading volume for filtration of challenging samples
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)