Skip to Content
Merck
All Photos(1)

Documents

MAB846

Sigma-Aldrich

Anti-Mumps Blend Antibody, fusion protein, clones 570-7A, 570-12G

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

570-12G, monoclonal
570-7A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

Specificity

Specific for mumps and has shown reactivity to the mumps ENDERS strain.

Immunogen

Epitope: fusion protein

Application

Anti-Mumps Blend Antibody, fusion protein, clones 570-7A, 570-12G is an antibody against Mumps Blend for use in ELISA, IF & WB.
EIA at 1:1,000 - 3,000.

Immunofluorescence at 1:1,000 - 3,000.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
In 0.02M phosphate buffer, 0.25 NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at +2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Vermeire et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 25(7), 907-907 (2018-12-07)
Mumps used to affect children between 2 and 15 years old. The mumps-measles-rubella (MMR) vaccine is available, with vaccine coverage rate of about 85% after two vaccine doses. Recently new mumps outbreaks have emerged in highly vaccinated populations; the causes for
Mumps and diabetes.
W P D LOGAN
Monthly bulletin of the Ministry of Health and the Public Health Laboratory Service null
Stephanie Franz et al.
Journal of virology, 91(18) (2017-07-01)
The mumps virus (MuV) small hydrophobic protein (SH) is a type I membrane protein expressed in infected cells. SH has been reported to interfere with innate immunity by inhibiting tumor necrosis factor alpha (TNF-α)-mediated apoptosis and NF-κB activation. To elucidate
J S Ellis et al.
Journal of clinical microbiology, 35(8), 2076-2082 (1997-08-01)
Multiple-target (multiplex) reverse transcription-PCR (RT-PCR) for detection, typing, and subtyping of the hemagglutinin gene of influenza type A (H3N2 and H1N1) and type B viruses was developed and applied prospectively to virological surveillance of influenza in England in the 1995-1996

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service