Skip to Content
Merck
All Photos(1)

Key Documents

WHA68832504

Whatman® GD/X syringe filters

regenerated cellulose membrane, pore size 0.45 μm, diam. 25 mm, non-sterile, pk of 1500 ea

Synonym(s):

Whatman filter, Z747629, filter syringe, syringe filter

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$5,130.00

$5,130.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$5,130.00

About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

$5,130.00


Check Cart for Availability

material

polypropylene housing
regenerated cellulose membrane

sterility

non-sterile

feature

glass fiber prefilter

packaging

pk of 1500 ea

manufacturer/tradename

Whatman 6883-2504
Whatman Article No. 28422289 (US reference)

diam.

25 mm

volume

≤25 mL

pore size

0.45 μm

fitting

female Luer inlet (lock)
male Luer outlet

Looking for similar products? Visit Product Comparison Guide

General description

Whatman GD/X non-sterile syringe filters with a regenerated cellulose (RC) membrane, suitable for hard-to-filter material with high solids. Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples. The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.

Features and Benefits

  • Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
  • Polyethersulfone (PES) for serum, plasma and tissue culture
  • Polyvinylidene difluoride (PVDF) for HPLC
  • Cellulose acetate for biological solutions
  • Borosilicate glass microfiber

Suitability

Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service