Key Documents
WHA420400
Whatman® plastic filter holders
Swin-Lok Holder, diam. 47 mm
Synonym(s):
Whatman filter, Whatman filters, Z742694
Select a Size
8 EA
$380.00
$380.00
Check Cart for Availability
Select a Size
Change View
8 EA
$380.00
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
$380.00
Check Cart for Availability
material
polycarbonate housing
sterility
non-sterile
feature
Swin-Lok Holder
packaging
pkg of 8 ea
manufacturer/tradename
Whatman 420400
Whatman Article No. 28419556 (US reference)
diam.
47 mm
General description
Filter holders designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
Features and Benefits
  • Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
  • All three holders will accommodate Nucleopore track-etched and cast membranes.
  • Syringe compatible.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)