Key Documents
WHA420100
Whatman® plastic filter holders
Pop-Top Holder, diam. 13 mm
Synonym(s):
Whatman filter, Whatman filters, Z742678
Select a Size
1 PKG
$159.00
$159.00
Available to ship on23 April 2025Details
Select a Size
Change View
1 PKG
$159.00
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
$159.00
Available to ship on23 April 2025Details
material
polycarbonate housing
sterility
non-sterile
feature
Pop-Top Holder
packaging
pkg of 10 ea
manufacturer/tradename
Whatman 420100
Whatman Article No. 28419554 (US reference)
diam.
13 mm
General description
Filter holders designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
Features and Benefits
  • Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
  • All three holders will accommodate Nucleopore track-etched and cast membranes.
  • Syringe compatible.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)