WHA2631990
Whatman® indicator papers
pH indicators, integral comparison strip, 9.5 to 12.0 range
Synonym(s):
Z753335
Select a Size
$49.90
Select a Size
About This Item
$49.90
Recommended Products
packaging
pk of 1
manufacturer/tradename
Whatman 2631-990
Whatman Article No. 28418931 (US reference)
L × W
11 mm × 100 mm
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
General description
pH Indicator Strips type CF (color bonded dye system)
Individual plastic support strips carry four different segments of dye-impregnated pH indicator papers. The resulting combination of color differences gives an extremely clear and accurate visual pH value. All the dyes are chemically bonded to the paper and cannot leach into solution, avoiding sample contamination and resultant anomalous pH readings.
pH Indicator Strips type CS (integral comparison chart)
Each pH test strip has a central segment of indicator dye and, printed alongside, eight or more different color segments marked with corresponding pH values for matching purposes. The pH test value can be read by direct comparison of the pH test strip color against the integrated color bars. pH indicator strips with integrated comparison bars are excellent for colored solutions so that any changes in the color of the paper stock are automatically disregarded.
pH Indicator Strip Dispensers type TC (triple color band)
The pH test strip has three separate indicator dye color bands. It comes as a roll in a convenient dispenser that helps to protect the indicator paper from fumes or aerosols that may change the color of the paper. The individual combination of color change resulting from each test is compared with the color-coded comparison chart printed on the dispenser for fast and accurate pH testing.
pH Indicator Strip Dispensers type SR (standard range)
A full range including some narrow ranges of pH indicator papers are available in a convenient dispenser.
pH Indicator Strip books
The book format is convenient for quick and economical pH testing, particularly for educational use.
Features and Benefits
- Instant pH readings
- Accuracy over a wide range of routine pH tests
- Convenience and portability for field use
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service