WHA1810110
Whatman® borosilicate glass TCLP testing filters 0.7 μm pore size
white, acid-treated, low metal, 50 ea, 110 mm diam
Synonym(s):
Whatman filtration, Z740799, metal filtration, tclp circles
Select a Size
$216.00
Select a Size
About This Item
$216.00
Recommended Products
Product Name
Whatman® acid treated low metal TCLP circles, diam. 110 mm
material
borosilicate glass fiber (BGF) membrane (acid treated low metal)
sterility
non-sterile
packaging
pkg of 50 ea
manufacturer/tradename
Whatman 1810-110
Whatman Article No. 28418302 (US reference)
parameter
60 mL/min speed
diam.
110 mm
pore size
0.7 μm
basis weight
75 g/m2
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Toxicity Characteristic Leaching Procedure (TCLP) is an analytical test designed to determine the leaching potential in a landfill for hazardous organic and inorganic contaminants that could potentially migrate into groundwater, threatening drinking water sources.
Used for EPA Method 1311
The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6 μm to 0.8 μm.
Features and Benefits
- CLP-compliant test filters from Cytiva come in six filter sizes for accurate landfill leachate modeling and sample analysis:
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service