Skip to Content
Merck
All Photos(1)

Key Documents

W526509

Sigma-Aldrich

Coumarin

≥98%

Synonym(s):

1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Beilstein:
383644
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

Halal

vapor pressure

0.01 mmHg ( 47 °C)

Assay

≥98%

origin

China origin

bp

298 °C (lit.)

mp

68-73 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous; sweet

SMILES string

O=C1Oc2ccccc2C=C1

InChI

1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H

InChI key

ZYGHJZDHTFUPRJ-UHFFFAOYSA-N

General description

Coumarin is an aroma compound responsible for the characteristic odor of hay.[1] It is one of the major volatile compounds found in the aerial parts of bastard balm.[2]

Application

It can be used an industrial odorant or odor masking agent.[3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

323.6 °F - closed cup

Flash Point(C)

162 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Volatile components of whole and different plant parts of bastard balm (Melittis melissophyllum L., Lamiaceae) collected in Central Italy and Slovakia.
Maggi F, et al.
Chemistry and Biodiversity, 8(11), 2057-2079 (2011)
Coumarin
Boisde PM and Meuly WC
Kirk-Othmer Encyclopedia of Chemical Technology (2000)
The pharmacology, metabolism, analysis, and applications of coumarin and coumarin-related compounds.
D Egan et al.
Drug metabolism reviews, 22(5), 503-529 (1990-01-01)
Shigehiro Sumiya et al.
The journal of physical chemistry. A, 117(7), 1474-1482 (2013-01-25)
A coumarin-amide-dipicolylamine linkage (L) was synthesized and used as a fluorescent receptor for metal cations in water. The receptor dissolved in water with neutral pH shows almost no fluorescence due to the photoinduced electron transfer (PET) from the amide and
Shota Morimoto et al.
Chemical communications (Cambridge, England), 49(18), 1811-1813 (2013-01-26)
A photo-switchable fluorescent flagging approach has been developed to identify photoaffinity-labeled peptides in target protein. Upon photochemical release of the ligand, the protein was newly modified with a coumarin in place of the previously attached biotin. It allowed us to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service