Skip to Content
Merck
All Photos(1)

Key Documents

8.22316

Sigma-Aldrich

Coumarin

for synthesis

Synonym(s):

Coumarin, 5,6-Benzo-2-pyrone

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$90.20
1 KG
$281.00

$90.20


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
$90.20
1 KG
$281.00

About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-086-7
NACRES:
NA.22

$90.20


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

1.3 hPa ( 106 °C)

Quality Level

Assay

≥99.0% (HPLC)

form

crystals

potency

293 mg/kg LD50, oral (Rat)

mp

68-71 °C

transition temp

flash point 162 °C

density

0.935 g/cm3

storage temp.

2-30°C

SMILES string

O1c2c(cccc2)C=CC1=O

InChI

1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H

InChI key

ZYGHJZDHTFUPRJ-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 68 °C
Melting range (upper value): ≤ 71 °C
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

323.6 °F - closed cup

Flash Point(C)

162 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service