Skip to Content
Merck
All Photos(1)

Documents

W515507

Sigma-Aldrich

2,5-Dimethylthiophene

≥98%

Synonym(s):

NSC 60689

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8S
CAS Number:
Molecular Weight:
112.19
Beilstein:
106450
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.064
NACRES:
NA.21

biological source

synthetic

grade

Halal
Kosher

Assay

≥98%

refractive index

n20/D 1.512 (lit.)

bp

134 °C/740 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

SMILES string

Cc1ccc(C)s1

InChI

1S/C6H8S/c1-5-3-4-6(2)7-5/h3-4H,1-2H3

InChI key

GWQOOADXMVQEFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dimethylthiophene is a volatile flavoring compound that has been identified in the essential oil of onion. It is reported to be one of the sulfur-containing compounds formed via Maillard reaction/Strecker degradation of cysteine with furaneol.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of sulfur-containing flavor compounds from reactions of furaneol and cysteine, glutathione, hydrogen sulfide, and alanine/hydrogen sulfide.
Zheng Y, et al.
Journal of Agricultural and Food Chemistry, 45(3), 894-897 (1997)
Volatile flavor compounds from onion.
Boelens M, et al.
Journal of Agricultural and Food Chemistry, 19(5), 984-991 (1971)
Restituto Tocmo et al.
Journal of agricultural and food chemistry, 62(23), 5296-5304 (2014-05-21)
There is a growing account of the health benefits of H2S as an endogenous cell-signaling molecule. H2S from organic polysulfides, in particular, is increasingly gaining attention for their beneficial effects to cardiovascular health. Here, we studied shallot as a potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service