Skip to Content
Merck
All Photos(1)

Key Documents

225797

Sigma-Aldrich

3-Acetyl-2,5-dimethylthiophene

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$86.30

$86.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$86.30

About This Item

Empirical Formula (Hill Notation):
C8H10OS
CAS Number:
Molecular Weight:
154.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.544 (lit.)

bp

105-108 °C/15 mmHg (lit.)

density

1.086 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1cc(C)sc1C

InChI

1S/C8H10OS/c1-5-4-8(6(2)9)7(3)10-5/h4H,1-3H3

InChI key

PUSJAEJRDNPYKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Acetyl-2,5-dimethylthiophene was used in the synthesis of heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone and 3-acetyl-2,5- dimethylthiophene semicarbazone[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krishna Sharma et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(1), 422-427 (2009-12-08)
Reactions of 3-acetyl-2,5-dimethylthiophene with thiosemicarbazide and semicarbazide hydrochloride resulted in the formation of new heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone (C(9)H(13)N(3)OS(2) or L(1)H) and 3-acetyl-2,5- dimethylthiophene semicarbazone (C(9)H(13)N(3)OS or L(2)H), respectively. The Pd(II) and Pt(II) complexes have been synthesized by mixing metal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service