Skip to Content
Merck
All Photos(1)

Key Documents

W385409

Sigma-Aldrich

3-Mercapto-3-methylbutan-1-ol

≥98%, FG

Synonym(s):

3-Mercapto-3-methylbutanol

Sign Into View Organizational & Contract Pricing

Select a Size

W385409-SAMPLE-K
$333.00
25 G
$735.00
100 G
$2,680.00

$333.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W385409-SAMPLE-K
$333.00
25 G
$735.00
100 G
$2,680.00

About This Item

Empirical Formula (Hill Notation):
C5H12OS
CAS Number:
Molecular Weight:
120.21
FEMA Number:
3854
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.137
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

$333.00


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

refractive index

n20/D 1.480

density

0.985 g/mL at 20 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty

storage temp.

2-8°C

SMILES string

CC(C)(S)CCO

InChI

1S/C5H12OS/c1-5(2,7)3-4-6/h6-7H,3-4H2,1-2H3

InChI key

GBCGIJAYTBMFHI-UHFFFAOYSA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service