Skip to Content
Merck
All Photos(1)

Key Documents

W314811

Sigma-Aldrich

Ethyl 2-trans-4-cis-decadienoate

natural, ≥95%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W314811-SAMPLE-K
$97.80
25 G
$380.00
100 G
$1,160.00
1 KG
$9,080.00

$97.80


Estimated to ship on30 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W314811-SAMPLE-K
$97.80
25 G
$380.00
100 G
$1,160.00
1 KG
$9,080.00

About This Item

Linear Formula:
CH3(CH2)4CH=CHCH=CHCO2C2H5
CAS Number:
Molecular Weight:
196.29
FEMA Number:
3148
Beilstein:
1724176
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.260
NACRES:
NA.21
Organoleptic:
apple; fatty; green; pear; sweet
grade:
FG
Fragrance grade
Halal
Kosher
natural
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

$97.80


Estimated to ship on30 March 2025


Request a Bulk OrderRequest more information

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥95%

refractive index

n20/D 1.486 (lit.)

bp

70-72 °C/0.05 mmHg (lit.)

density

0.905 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; fatty; green; pear; sweet

SMILES string

[H]\C(CCCCC)=C([H])\C([H])=C\C(=O)OCC

InChI

1S/C12H20O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h8-11H,3-7H2,1-2H3/b9-8-,11-10+

InChI key

OPCRGEVPIBLWAY-QNRZBPGKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Natural occurrence: Apple, pear, grape, pear brandy and quince.

Application

Potential applications include the profile enhancement of guava, melon, pear, apple, banana, kiwi, grape, golden delicious apple, fruit cocktail juices, fruit nectar, mamey and other tropical backgrounds.

Biochem/physiol Actions

Odor at 1.0%
Taste at 5 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30(5), 49-49 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service