Skip to Content
Merck
All Photos(1)

Documents

W280220

Sigma-Aldrich

2-Octanone

natural, 98%, FG

Synonym(s):

Hexyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5COCH3
CAS Number:
Molecular Weight:
128.21
FEMA Number:
2802
Beilstein:
635843
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.019
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

98%

refractive index

n20/D 1.416 (lit.)

bp

173 °C (lit.)

mp

−16 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

woody; earthy

SMILES string

CCCCCCC(C)=O

InChI

1S/C8H16O/c1-3-4-5-6-7-8(2)9/h3-7H2,1-2H3

InChI key

ZPVFWPFBNIEHGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shane Avison et al.
Journal of agricultural and food chemistry, 63(28), 6313-6318 (2015-06-23)
A new methodology is presented to measure water-air partition coefficients (Henry's constants) of volatiles, using APCI-MS. Significant advantages over other Henry's constant determination methods include the short measurement and sample preparation time and the possibility for simultaneous measurement of multiple
Paweł Mochalski et al.
Cell biochemistry and biophysics, 71(1), 323-329 (2014-08-16)
Gas chromatography with mass spectrometric detection combined with head-space needle trap extraction as the pre-concentration technique was applied to identify and quantify volatile organic compounds released or metabolised by human umbilical vein endothelial cells. Amongst the consumed species there were
Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric
Éder Costa Oliveira et al.
Talanta, 127, 59-67 (2014-06-11)
Hollow fiber liquid phase microextraction (HFLPME) and solid phase microextraction (SPME) methods for pre-concentration of contaminants (toluene, benzophenone, tetracosane and chloroform) in food simulants were investigated. For HFLPME 1-heptanol, 2-octanone and dibutyl-ether were studied as extracting solvents. Analysis by gas
Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service