Merck
  • Novel Methodology for Measuring Temperature-Dependent Henry's Constants of Flavor Molecules.
Novel Methodology for Measuring Temperature-Dependent Henry's Constants of Flavor Molecules.
Journal of agricultural and food chemistry (2015-06-23)
Shane Avison, Kitty van Gruijthuijsen, Mirela Pascu, Alan Parker, Igor Bodnár
ABSTRACT
A new methodology is presented to measure water-air partition coefficients (Henry's constants) of volatiles, using APCI-MS. Significant advantages over other Henry's constant determination methods include the short measurement and sample preparation time and the possibility for simultaneous measurement of multiple volatiles. The methodology is validated by obtaining good agreement with reliable literature values for a series of 2-ketones. The methodology is further explored for eight key volatiles typically found in citrus fruits, including the temperature dependence of the Henry's constant. Using these data can improve estimates of flavor losses during processing and volatile release during consumption.
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Octanone, ≥98%, FG
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
2-Octanone, reagent grade, 98%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
2-Heptanone, natural, 98%, FG
Sigma-Aldrich
2-Pentanone, ≥98%, FCC, FG