Skip to Content
Merck
All Photos(1)

Key Documents

W259411

Sigma-Aldrich

α-Ionone

natural, ≥86%

Synonym(s):

4-(2,6,6-Trimethyl-2-cyclohexenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
FEMA Number:
2594
Beilstein:
3197885
EC Number:
Council of Europe no.:
141
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.007
NACRES:
NA.21

grade

Halal
Kosher
natural

Quality Level

reg. compliance

FDA 21 CFR 117

Assay

≥86%

refractive index

n20/D 1.498 (lit.)

bp

259-263 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral

SMILES string

CC(=O)\C=C\C1C(C)=CCCC1(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h6-8,12H,5,9H2,1-4H3/b8-7+

InChI key

UZFLPKAIBPNNCA-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darunee Soorukram et al.
Organic letters, 6(14), 2409-2411 (2004-07-02)
[reaction: see text] The allylic substitution of sterically hindered (2-iodocycloalkyl)phosphates proceeds with complete anti S(N)2' stereoselectivity with mixed diorganozincs of the type RZnCH(2)SiMe(3) in the presence of CuCN.2LiCl. Only the group R of the copper-zinc reagent is transferred in the
Hao Cai et al.
Molecules (Basel, Switzerland), 18(2), 1368-1382 (2013-01-26)
Flos Lonicerae Japonicae (FLJ) is a popular herb used for many centuries in Traditional Chinese Medicine as a treatment of fever and inflammation. Non-fumigated processing of FLJ has been the traditional approach used in post-harvest preparation of the commodity for
Yonghong Wu et al.
Environmental microbiology, 13(3), 604-615 (2010-11-09)
Periphyton biofilms are natural mixtures comprised of photoautotrophic and heterotrophic complex microorganisms. In this work, the inhibition effects of periphyton biofilms on cyanobacterial blooms were studied in pilot and field trials. Results show that the cyanobacterial species responsible for the
Sabine Sewenig et al.
Journal of agricultural and food chemistry, 53(4), 838-844 (2005-02-17)
A new coupling system of GC-GC, connected via a Multi Column Switching Device MCS2 for measuring isotope ratios, is introduced. By means of several standard substances the precise and accurate measurement of isotopic values is proved. First applications concerning the
Ufuk Ozgen et al.
Molecules (Basel, Switzerland), 15(4), 2593-2599 (2010-04-30)
Sonchus erzincanicus (Asteraceae) is an endemic species in Turkey, where six Sonchus species grow. In this study, a phytochemical study was performed on the aerial parts of the plant. The study describes the isolation and structure elucidation of five flavonoids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service