Skip to Content
Merck
All Photos(1)

Documents

W209805

Sigma-Aldrich

Anisyl acetate

≥97%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2C6H4-4-(OCH3)
CAS Number:
Molecular Weight:
180.20
FEMA Number:
2098
EC Number:
Council of Europe no.:
209
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.019
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.513 (lit.)

bp

137-139 °C/12 mmHg (lit.)

density

1.107 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

anise; cherry; sweet; vanilla

SMILES string

COc1ccc(COC(C)=O)cc1

InChI

1S/C10H12O3/c1-8(11)13-7-9-3-5-10(12-2)6-4-9/h3-6H,7H2,1-2H3

InChI key

HFNGYHHRRMSKEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 30 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mosciano, G.
Perfumer & Flavorist, 24, 49-49 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service