Skip to Content
Merck
All Photos(1)

Key Documents

W215805

Sigma-Aldrich

Isoborneol

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2158
EC Number:
Council of Europe no.:
2020
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.059
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

description

Stabilized

Assay

≥95%

contains

contains BHT, synthetic as stabilizer

mp

212-214 °C (subl.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

woody; camphoraceous; balsamic

SMILES string

[H][C@@]12CC[C@@](C)([C@H](O)C1)C2(C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8-,10+/m1/s1

InChI key

DTGKSKDOIYIVQL-MRTMQBJTSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S182-S184 (2008-07-22)
A toxicologic and dermatologic review of isoborneol when used as a fragrance ingredient is presented.
Eva Horváthová et al.
Mutagenesis, 27(5), 581-588 (2012-05-01)
Experimental evidences suggest that most essential oils possess a wide range of biological and pharmacological activities that may protect tissues against oxidative damage. In this study, we investigated DNA-protective effect of borneol, a component of many essential oils, against oxidative
Jing-yi Zhao et al.
Journal of Zhejiang University. Science. B, 13(12), 990-996 (2012-12-12)
Borneol, a monoterpenoid alcohol, is used widely, particularly in combined formulas for preventing and curing cardiovascular and cerebrovascular diseases in traditional Chinese medicine. In order to understand the blood and brain pharmacokinetics after intravenous, intranasal, or oral administration and to
Xiao-Hong Wei et al.
Microcirculation (New York, N.Y. : 1994), 20(1), 17-29 (2012-08-24)
The present study was designed to evaluate whether CP was beneficial in alleviating myocardial fibrosis following I/R injury. Sprague-Dawley rats were subjected to 30 minutes occlusion of the LADCA, followed by reperfusion. CP (0.4 or 0.8 g/kg) was daily administered starting from
Xuxiao Lv et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 37(7), 878-881 (2012-07-17)
Malignant tumor, epilepsy, dementia, cerebral ischemia and other brain diseases have very high rates of disability and mortality. Currently, many drugs are developed to treat such diseases and the effect is obviously. But they can not achieve the purpose to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service