Skip to Content
Merck
All Photos(1)

Key Documents

461946

Sigma-Aldrich

Poly(2-vinylnaphthalene)

average Mw ~175,000

Synonym(s):

2-Vinylnaphthalene homopolymer, 2-Vinylnaphthalene polymer, Poly(β-vinylnaphthalene)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$405.00

$405.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$405.00

About This Item

Linear Formula:
[CH2CH(C10H7)]n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$405.00


Check Cart for Availability

Request a Bulk Order

mol wt

average Mw ~175,000

transition temp

Tg (DSC) 135 °C (onset)

SMILES string

c21c(ccc(c2)C=C)cccc1

InChI

1S/C12H10/c1-2-10-7-8-11-5-3-4-6-12(11)9-10/h2-9H,1H2

InChI key

KXYAVSFOJVUIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Molecular Weight Dependence of Triplet-Triplet Processes in Poly(2-vinylnaphthalene): This research examines the effects of molecular weight on the triplet-triplet processes in poly(2-vinylnaphthalene) at low temperatures, providing insights into the photophysical properties of this polymer (N Eliezer, NBS Special Publication 526).
  • Atom Transfer Radical Polymerization in the Solid-State: This paper discusses the synthesis of poly(2-vinylnaphthalene) via solid-state atom transfer radical polymerization, highlighting a novel approach to polymer synthesis that reduces solvent use (HY Cho, CW Bielawski, Angewandte Chemie, 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W J Litchfield et al.
Clinical chemistry, 30(9), 1489-1493 (1984-09-01)
Recent innovations in particle design have led to the development of highly sensitive and reproducible immunoassay methods for the Du Pont aca discrete clinical analyzer. Key advances include the synthesis and use of particles less than 1 micron in diameter
Fengyang Wang et al.
Macromolecular rapid communications, 33(10), 933-937 (2012-03-01)
A straightforward strategy for assembling polymeric dimers from amphiphilic nanoparticles is reported. Amphiphilic polymeric nanoparticles with a mixed-shell of PEO/P2VN blocks and a flexible core of PAA blocks are fabricated by a non-covalent crosslinking method. Uniform polymeric dimers are efficiently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service