Key Documents
TEM-LC400-AU-100
Sigma-Aldrich
Lacey Carbon Supported Gold Grid
size 400 mesh, hole size 100 μm , Average, box of 25
Select a Size
1 PKG
$198.00
$198.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$198.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$198.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 25
hole size
100 μm , Average
size
400 mesh
General description
Lacey Carbon Supported Grid: Gold Grid, 400 mesh. Average hole size is 100 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)