Key Documents
TEM-FFT200CU25
Sigma-Aldrich
Formvar Supported Copper Thin Bar Square Mesh
size 200 mesh, box of 25
Synonym(s):
FORMVAR SUPPORT FILM THIN BAR SQUARE 200 MESH CU GRIDS
Select a Size
1 PKG
$203.00
$203.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$203.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$203.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 25
size
200 mesh
General description
A 10nm thin film of pure formvar resin supported thin bar square mesh grids. Copper Grid, 200 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)