Key Documents
TEM-FF100-AU-UL
Sigma-Aldrich
Ultral-Thin Formvar Supported Gold Square Mesh
size 100 mesh, box of 50
Select a Size
1 PKG
$132.00
$132.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$132.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$132.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
100 mesh
General description
A 5-6 nm ultral-thin formvar film supported standard square mesh grids. Gold grid, 100 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)