Key Documents
TEM-FF150CU50
Sigma-Aldrich
Formvar Supported Copper Square Mesh
size 150 mesh, box of 50
Synonym(s):
FORMVAR FILM 150 MESH VU 50/ BX
Select a Size
1 PKG
$163.00
$163.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$163.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$163.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
150 mesh
General description
A 10 nm thin formvar film supported standard copper square mesh grids. Copper grid, 150 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)