Key Documents
TEM-7596410
Sigma-Aldrich
OMNI Probe Copper 4-Post Lift-Out Grid
pack of 100 (per vial)
Synonym(s):
Omni Grids - Copper 4-Post Lift-Out Grids 100/pk
Select a Size
1 PKG
$283.00
$283.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$283.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$283.00
Check Cart for Availability
Request a Bulk Order
packaging
pack of 100 (per vial)
General description
4 post copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations, two with vertical bar attachment surfaces and two with "V" shaped alighment surfaces. Sides have lower profile for easier access to outermost posts. 3mm diameter.
Packaging
Packaged in plastic vials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)