Key Documents
TEM-CF200-NI-TH
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
Thick, size 200 mesh, box of 50
Select a Size
1 PKG
$152.00
$152.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$152.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$152.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
200 mesh
General description
A 10 nm thick film of pure carbon supported standard squre mesh grids. Nickel grid, 200 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)