Key Documents
TEM-CF150-CU-ET
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
Extra Thick, size 150 mesh, box of 50
Select a Size
1 PKG
$165.00
$165.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$165.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$165.00
Check Cart for Availability
Request a Bulk Order
packaging
box of 50
size
150 mesh
General description
A 20-30 nm thick film of pure carbon supported standard squre mesh grids. Copper grid, 150 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)