Key Documents
TEM-75965-01
Sigma-Aldrich
Omni Lift-Out Grid Storage Box
pkg of 1 ea
Select a Size
1 PKG
$34.70
$34.70
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$34.70
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$34.70
Check Cart for Availability
Request a Bulk Order
packaging
pkg of 1 ea
General description
Storage box for 100 standard or haft grids—3 mm diameter –TEM grids. Box comes complete with base, lid and clips.
Choose from one of the most recent versions:
Certificates of Analysis (COA)