Key Documents
TEM-7596407
Sigma-Aldrich
OMNI Probe Copper 3-Post Lift-Out Grid
pack of 100 (per vial)
Synonym(s):
Omniprobe Lift-Out Grid, 3-Post, Side Access, CU
Select a Size
1 PKG
$265.00
$265.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$265.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$265.00
Check Cart for Availability
Request a Bulk Order
packaging
pack of 100 (per vial)
General description
3 post copper lift-out grids, 35 micron thick with 1 edge lower for easy access.
Packaging
Packaged in glass vials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)