Key Documents
Z362727
Sigma-Aldrich
Nalgene® polycarbonate Erlenmeyer flask
capacity 125 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
Select a Size
12 EA
$414.00
$414.00
Available to ship on14 April 2025Details
Select a Size
Change View
12 EA
$414.00
About This Item
UNSPSC Code:
41121804
eCl@ss:
32040501
NACRES:
NB.43
$414.00
Available to ship on14 April 2025Details
material
flat bottom
polycarbonate
wide-neck
Joint
joint stopper5
sterility
non-sterile
feature
closure type Stoppers
without cap
packaging
pack of 12
case of 24
manufacturer/tradename
Nalgene 4103-0125
capacity
125 mL
General description
Autoclavable; however, repeated autoclaving may cause some loss of mechanical strength.
Other Notes
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)