Key Documents
TEM-71166-10
Sigma-Aldrich
Cryo Grid Box
round, w/lid
Select a Size
1 PKG
$21.60
$21.60
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 PKG
$21.60
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$21.60
Check Cart for Availability
Request a Bulk Order
General description
This specimen grid boxe is for storing or transferring cryogenic TEM specimen grids. It features four diamond shaped slots, non-static clear covershield held in place with stainless steel screw, which is tapped in the center of the box. Box fits the FEI Vitrobot, Gatan 626, Gatan 3500, comes with lid, round, this is one unit of cryo grid box.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. How many grid boxes are contained in a package?
    1 answer
    1. This product includes one Cryo Grid Box per package.
      Helpful?