Skip to Content
Merck
All Photos(1)

Key Documents

593656

Sigma-Aldrich

4-tert-Butylbenzenethiol

97%

Synonym(s):

4-tert-Butylthiophenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$168.00
25 G
$452.00

$168.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$168.00
25 G
$452.00

About This Item

Linear Formula:
(CH3)3CC6H4SH
CAS Number:
Molecular Weight:
166.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$168.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.5480 (lit.)

bp

238 °C (lit.)

density

0.964 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccc(S)cc1

InChI

1S/C10H14S/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11H,1-3H3

InChI key

GNXBFFHXJDZGEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-tert-Butylbenzenethiol can be used to synthesize Pd–thiolate complex by coordinating with Pd2+.The reduction of this complex forms ultrasmall palladium nanoclusters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongbo Song et al.
Nature communications, 11(1), 478-478 (2020-01-26)
Gold-copper alloys have rich forms. Here we report an atomically resolved [Au52Cu72(p-MBT)55]+Cl- nanoalloy (p-MBT = SPh-p-CH3). This nanoalloy exhibits unusual structural patterns. First, two Cu atoms are located in the inner 7-atom decahedral kernel (M7, M = Au/Cu). The M7 kernel is then enclosed
Gaya N Andrew et al.
Chemistry, an Asian journal, 15(23), 4077-4081 (2020-10-14)
We report the single-crystal synthesis of a chlorine-centered bimetallic cluster, Cl@Ag22 Au6 (4-TBBT)28 (PPh4 ), which bears a quatrefoil-structured Cl@Ag22 (SR)16 core studded by six Au(SR)2 staples showing a quasi Td symmetry. This cluster bears 28 metal atoms and 28
Masaaki Yusa et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 16(7), 1043-1048 (2017-05-10)
Under the irradiation of red light (690 nm), quinones were converted to hydroquinones by thiols in the presence of metallophthalocyanines. The reaction proceeded via the charge separation between the triplet state of phthalocyanine and the quinone. The product determining step

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service