Key Documents
SYNM431003
Synthware all-glass vacuum/inert gas manifold with hollow high vacuum stopcocks
number of ports, 3, Hose Connections: Front-left, Rear-left-right
Synonym(s):
Synthware M431003, vacuum manifold, vacuum unit
Select a Size
1 EA
$931.00
$931.00
Check Cart for Availability
Select a Size
Change View
1 EA
$931.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$931.00
Check Cart for Availability
feature
number of ports 3
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M431003
General description
With 4 mm double oblique, high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. Beams are 22 mm O.D. with front-left and rear-left-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)