Key Documents
SYNM290004
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 4, Hose Connections: Front-left-right, Rear-right
Synonym(s):
Glass vacuum manifold, High vacuum manifold, Inert gas system, Synthware M290004, vacuum manifold, vacuum unit
Select a Size
1 EA
$1,210.00
$1,210.00
In StockDetails
Select a Size
Change View
1 EA
$1,210.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$1,210.00
In StockDetails
feature
number of ports 4
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M290004
General description
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-right 10 mm hose connections. Distance between stopcocks is75 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)